Learn More
Grainger Mounting Bracket Steel Swivel
Catalog No. 502765705
Encompass_Preferred
The COXREELS Mounting Bracket is made of steel and is designed for use with Coxreels hose reels. It has a black powder coat finish and includes mounting hardware. The bracket has pre-drilled holes for easy installation and is compatible with most Coxreels models. This heavy duty bracket securely mounts the hose reel to walls or ceilings. It measures 3 inches wide x 4 inches tall x 3/8 inches thick. The bracket is made in the USA and comes with a 5 year warranty from the manufacturer.
Catalog No. 50-276-5705 Supplier Grainger Supplier No. 16X562
The COXREELS Mounting Bracket is made of steel and is designed for use with Coxreels hose reels. It has a black powder coat finish and includes mounting hardware. The bracket has pre-drilled holes for easy installation and is compatible with most Coxreels models. This heavy duty bracket securely mounts the hose reel to walls or ceilings. It measures 3 inches wide x 4 inches tall x 3/8 inches thick. The bracket is made in the USA and comes with a 5 year warranty from the manufacturer.