Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Easy-Load Pump Head Mounting Hardware, Mounts Three Pump Head, Stainless Steel; 2/Set
Catalog No. NC9783681
Encompass
This mounting hardware is designed to make the Masterflex L/S series pump head flexible for a wide range of applications. Choose from different material or increase the number of pump heads on a L/S drive with mounting hardware. This hardware is compatible with the Masterflex L/S series peristaltic drives.
Catalog No. NC9783681 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0701308
This mounting hardware is designed to make the Masterflex L/S series pump head flexible for a wide range of applications. Choose from different material or increase the number of pump heads on a L/S drive with mounting hardware. This hardware is compatible with the Masterflex L/S series peristaltic drives.