Learn More
Hach Company Mounting Hardware Tee SS
Catalog No. 5000207156 Shop All Hach Company Products
Encompass
Mounting Hardware Tee-316 SS- 75 Each tee is a standard 3/4-inch tee with 3/4-inch NPT threads on all three openings
Catalog No. 50-002-07156 Supplier Hach Company Supplier No. MH313N3NZ
Mounting Hardware Tee-316 SS- 75 Each tee is a standard 3/4-inch tee with 3/4-inch NPT threads on all three openings