Learn More
Hach Company Mounting Kit Dig. pH Sensor
Supplier:  Hach Company 6136400
Encompass
Immersion Mounting Kit for Digital pH SensorCPVC Immersion Mount Includes 1-Inch Diameter By 4 Ft Long Pipe And 1-Inch X 1-Inch For Use With Hach Digital Phd Sensors
Catalog No. 50-002-07179