Learn More
Andwin Scientific MOUNTING MEDIA 16OZ
SDP
Catalog No. NC9016679
Encompass_Preferred
The Tissue-Tek Glas Mounting Medium is compatible to use with the Glas Coverslipper to maximize performance, generating clean, bubble-free slides. This product can be opened and put directly onto the mounting medium area of the coverslipper without the need of pouring.
Catalog No. NC9016679 Supplier Andwin Scientific Supplier No. 6419
The Tissue-Tek Glas Mounting Medium is compatible to use with the Glas Coverslipper to maximize performance, generating clean, bubble-free slides. This product can be opened and put directly onto the mounting medium area of the coverslipper without the need of pouring.