Learn More
Abcam Mounting Medium for IHC - Aqueous, 125ML
Catalog No. NC1275930
Encompass_Preferred
Aqueous mounting medium for IHC is designed for use with chromogenic tissue and cell staining. - Works with both aqueous and non-aqueous chromogens (e.g., Fast-Red, AEC, DAB). - Forms a dry, even surface for direct visualization without glass coverslip
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1275930 Supplier Abcam Supplier No. AB64230125ML
Aqueous mounting medium for IHC is designed for use with chromogenic tissue and cell staining. - Works with both aqueous and non-aqueous chromogens (e.g., Fast-Red, AEC, DAB). - Forms a dry, even surface for direct visualization without glass coverslip
The product is subject to the following: Abcam Restricted Use Statement