Learn More
Ibidi USA Mounting Medium, 15 ml
SDP
Catalog No. 50305707
Encompass
ibidi Mounting Medium is a glycerol-based, non-hardening mounting medium optimized for fluorescence microscopy. The dropper bottle is ideal for exact dispensing into channels or open wells such as in the ibidi µ-Slides and µ-Dishes. 15 ml
This item is not returnable. View return policy
Catalog No. 50-305-707 Supplier Ibidi USA Supplier No. 50001
ibidi Mounting Medium is a glycerol-based, non-hardening mounting medium optimized for fluorescence microscopy. The dropper bottle is ideal for exact dispensing into channels or open wells such as in the ibidi µ-Slides and µ-Dishes. 15 ml