Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mounting Medium, Synth, Vion Biosciences
SCI_ED

Catalog No. S02515 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02515 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to Ethylbenzene, which is known to the State of California to cause cancer.
Catalog No. S02515 Supplier Aldon Corporation Supplier No. VNBB0020500ML
Add to Cart
Add to Cart
WARNING: This product can expose you to Ethylbenzene, which is known to the State of California to cause cancer.

This synthetic mounting medium is an alternative to natural balsam canada resin in xylene that can be used as a mounting medium in preparing permanent sample slides.

  • This synthetic mounting medium is an alternative to natural balsam canada resin in xylene that can be used as a mounting medium in preparing permanent sample slides.

Specifications

Applications Slide Mounting Media
Aqueous or Permanent Permanent
Volume (English) 500 mL
Quantity 500 mL
Product Type Mounting Medium
Packaging Amber Glass
Content And Storage Store in a cool, dry, well-ventilated area away from incompatible substances. Keep away from ignition sources.
Product Line Vion Histology
Reagent Type Mounting Solution
Volume (Metric) 500 mL
Description Balsam Canada, Synthetic in Xylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.