Learn More
Electron Microscopy Sciences EUKITT Neo Special Mounting Medium 100 ML
SDP
Catalog No. 501928504
Encompass_Preferred
EUKITT Neo Special is a xylene free product and is characterized by faster curing and slightly higher viscosity than EUKITT Neo. Users of EUKITT Classic will appreciate the xylene free replacement. The product retains crystal-clear optics, low fluorescence, favorable refractive index, good fluidity and low shrinkage properties. The preparations are stable without cracking and discoloration.
EUKITT Neo Special is colorless and does not change shape, structure or color of the material to be examined. The use of sustainable, renewable raw material as a solvent completes the modern formulation approach.
The pleasant odor facilitates working in the laboratory. Supplied in wide neck, unbreakable aluminum bottles.
Catalog No. 50-192-8504 Supplier Electron Microscopy Sciences Supplier No. 1532025
EUKITT Neo Special is a xylene free product and is characterized by faster curing and slightly higher viscosity than EUKITT Neo. Users of EUKITT Classic will appreciate the xylene free replacement. The product retains crystal-clear optics, low fluorescence, favorable refractive index, good fluidity and low shrinkage properties. The preparations are stable without cracking and discoloration.
EUKITT Neo Special is colorless and does not change shape, structure or color of the material to be examined. The use of sustainable, renewable raw material as a solvent completes the modern formulation approach.
The pleasant odor facilitates working in the laboratory. Supplied in wide neck, unbreakable aluminum bottles.