Learn More
Enzo Life Sciences SEEBRIGHT Mounting Medium with DAPI  (100tests)
SDP
Catalog No. 502013773
Encompass_Preferred
SEEBRIGHT Mounting Medium with DAPI is an antifade reagent that minimizes photobleaching. Use of the mounting medium significantly extends the lifetime of fluorescent signals. The inclusion of DAPI allows for staining of the nucleus. Quantity: Each vial of SEEBRIGHT Mounting Medium with DAPI contains 200 µL sufficient for 20 tests (ENZ-GEN420-0020) or 1000 µL sufficient for 100 tests (ENZ-GEN420-0100). Apply mounting medium to the sample area as desired. Long Term Storage: +4°C. Applications: IF, Fluorescence microscopy, Fluorescent detection FISH.
Catalog No. 50-201-3773 Supplier Enzo Life Sciences Supplier No. ENZGEN4200100
SEEBRIGHT Mounting Medium with DAPI is an antifade reagent that minimizes photobleaching. Use of the mounting medium significantly extends the lifetime of fluorescent signals. The inclusion of DAPI allows for staining of the nucleus. Quantity: Each vial of SEEBRIGHT Mounting Medium with DAPI contains 200 µL sufficient for 20 tests (ENZ-GEN420-0020) or 1000 µL sufficient for 100 tests (ENZ-GEN420-0100). Apply mounting medium to the sample area as desired. Long Term Storage: +4°C. Applications: IF, Fluorescence microscopy, Fluorescent detection FISH.