Learn More
Abcam Mounting Medium With DAPI - Aqueous, Fluoroshield, 100ML
Catalog No. 502585553
Encompass_Preferred
DAPI mounting medium (previously called Fluoroshield Mounting Medium With DAPI) is an aqueous anti-fade mounting medium designed to preserve fluorescence when imaging tissues and cell samples. - Cited in over 325 publications - Suitable for immunohistochemistry (IHC), immunocytochemistry/immunofluorescence (ICC/IF) and more - Contains 84% water, 0.0002% 4',6-Diamidino-2-phenylindole dihydrochloride
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5553 Supplier Abcam Supplier No. AB104139100ML
DAPI mounting medium (previously called Fluoroshield Mounting Medium With DAPI) is an aqueous anti-fade mounting medium designed to preserve fluorescence when imaging tissues and cell samples. - Cited in over 325 publications - Suitable for immunohistochemistry (IHC), immunocytochemistry/immunofluorescence (ICC/IF) and more - Contains 84% water, 0.0002% 4',6-Diamidino-2-phenylindole dihydrochloride
The product is subject to the following: Abcam Restricted Use Statement