Learn More
Electron Microscopy Sciences EUKITT Microfix Mounting Medium 100 ML
SDP
Catalog No. 501928506
Encompass_Preferred
MICROFIX follows the proven, xylene-based approach of O. Kindler ORSAtec. It is characterized by fast curing, crystal-clear optics, low fluorescence, ideal refractive index, good flowability and low shrinkage. It is colorless and does not change shape, structure or color of the material to be examined. These characteristics are already known from the classic EUKITT. What is new is the streamlined and optimized synthesis and filling process, and a still further optimized selection of our raw materials.
Catalog No. 50-192-8506 Supplier Electron Microscopy Sciences Supplier No. 1532035
MICROFIX follows the proven, xylene-based approach of O. Kindler ORSAtec. It is characterized by fast curing, crystal-clear optics, low fluorescence, ideal refractive index, good flowability and low shrinkage. It is colorless and does not change shape, structure or color of the material to be examined. These characteristics are already known from the classic EUKITT. What is new is the streamlined and optimized synthesis and filling process, and a still further optimized selection of our raw materials.