Learn More
Fritsch Milling And Sizing Inc Mounting Rack Cpl. for P-1/ P-13 Classic Line
SDP
Catalog No. 501534407
Encompass
For fine pre-crushing of sample material with a feed size of 95 mm down to 0.1 mm final fineness in a single process and for continuous operation with the Jaw Crushers PULVERISETTE 1 classic line
Fine pre-crushing from 95 mm down to 0.1 mm in a single process
A mounting rack with chute in combination with the Jaw Crusher PULVERISETTE 1 (Model I + II) classic line and the Disk Mill PULVERISETTE 13 classic line make it possible to grind 95 mm feed material down to a final material fineness of 100 µm in a single process. Continuous comminution is also possible.
Continuous operation
With the operation of the mounting rack and delivery chute, the jaw crushers PULVERISETTE 1 (Model I + II) classic line can be used for continuous operation.
Catalog No. 50-153-4407 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.5100.00
Add to Cart
Add to Cart
Request a Quote
For fine pre-crushing of sample material with a feed size of 95 mm down to 0.1 mm final fineness in a single process and for continuous operation with the Jaw Crushers PULVERISETTE 1 classic line
Fine pre-crushing from 95 mm down to 0.1 mm in a single process
A mounting rack with chute in combination with the Jaw Crusher PULVERISETTE 1 (Model I + II) classic line and the Disk Mill PULVERISETTE 13 classic line make it possible to grind 95 mm feed material down to a final material fineness of 100 µm in a single process. Continuous comminution is also possible.
Continuous operation
With the operation of the mounting rack and delivery chute, the jaw crushers PULVERISETTE 1 (Model I + II) classic line can be used for continuous operation.