Learn More
Electron Microscopy Sciences Mounting Wax 100 20 GR
SDP
Catalog No. 50997389
Encompass_Preferred
MWM100 Black Wax is designed for use as a masking material in jet polishing applications. As it is chemically inert, it is resistant to etching solutions such as hydrofluoric acid, perchloric acid and acetic acid. While it is resistant to many acids, it can be easily removed with hydrocarbon or chlorinated solvents. Each stick is 20 grams. Moderate Hardness and a Melting Point of 100 degree C. Dilutent: Hydrocarbon or Chlorinated solvents
Catalog No. 50-997-389 Supplier Electron Microscopy Sciences Supplier No. 50390100
MWM100 Black Wax is designed for use as a masking material in jet polishing applications. As it is chemically inert, it is resistant to etching solutions such as hydrofluoric acid, perchloric acid and acetic acid. While it is resistant to many acids, it can be easily removed with hydrocarbon or chlorinated solvents. Each stick is 20 grams. Moderate Hardness and a Melting Point of 100 degree C. Dilutent: Hydrocarbon or Chlorinated solvents