Learn More
Electron Microscopy Sciences Mounting Wax 52 100 GR
SDP
Supplier:  Electron Microscopy Sciences 5039052
Encompass_Preferred
MWS052 is a tan colored wax, which is translucent on thin films. Softness makes it most useful with abrasive slurry cutting of materials. Best suited to non-porous surfaces such as glass or polished metal with a reasonably large area. Easily soluble in a variety of solvents. Soft Hardness and a Melting Point of 52 degree C. Dilutent: Perchloroethylene 1" x 1" x 7.5" long (25 x 25 x 190 mm).
Catalog No. 50-997-390