Learn More
Electron Microscopy Sciences Mounting Wax 80 25 GR
SDP
Supplier:  Electron Microscopy Sciences 5039080
Encompass_Preferred
MWH080 is an amber colored wax, which is translucent in thin films. High hardness makes it ideal for sawing, ultrasonic cutting and lappingand polishing operations. Best suited to vitreous materials, polished metal surfaces, semiconductors and piezoelectric materials with large contacted areas. Chemical resistant. Hard hardness with a Melting Point of 80 degree C. Dilutent: Ethanol and Acetone
Catalog No. 50-997-392