Learn More
Thermo Scientific Viper Fingertight Fitting Systems | Stainless Steel | 1 EA
Catalog No. 501347813 Shop All Thermo Scientific Products
Encompass
Robust LC Performance, Ferrule-free tightening design for virtually zero-dead-volume connections, Operates at pressures up to 1310 bar, Suitable for temperatures up to 120C, Easy to Install, Fingertight design, Flexible stainless-steel 132 in OD, No tools required, Flexible, Compatible with valves and columns from virtually any manufacturer, Fits narrow connections such as 10-port valves, Available in different materials lengths and inner diameters, Recommended Use, For fluidic connections for analytical LC systems, Thermo Scientific Vanquish Core HPLC systems and Thermo Scientific UltiMate 3000 RS and SD systems are equipped with the Viper Fingertight Fitting System with stainless steel capillaries as a standard
Catalog No. 50-134-7813 Supplier Thermo Scientific Supplier No. 6040.2315
Robust LC Performance, Ferrule-free tightening design for virtually zero-dead-volume connections, Operates at pressures up to 1310 bar, Suitable for temperatures up to 120C, Easy to Install, Fingertight design, Flexible stainless-steel 132 in OD, No tools required, Flexible, Compatible with valves and columns from virtually any manufacturer, Fits narrow connections such as 10-port valves, Available in different materials lengths and inner diameters, Recommended Use, For fluidic connections for analytical LC systems, Thermo Scientific Vanquish Core HPLC systems and Thermo Scientific UltiMate 3000 RS and SD systems are equipped with the Viper Fingertight Fitting System with stainless steel capillaries as a standard