Learn More
Biomatik Corporation Mouse 11-Beta-Hydroxysteroid Dehydrogenase Type 1 (HSD11b1) ELISA Kit
Supplier:  Biomatik Corporation EKU09113
Encompass_Preferred
Mouse 11-Beta-Hydroxysteroid Dehydrogenase Type 1 (HSD11b1) ELISA Kit for detecting HSD11b1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50172. Gene ID: 15483. Target Synonyms: 11DH; 11-beta-HSD1; HDL; HSD11; HSD11B; HSD11L; SDR26C1; Corticosteroid 11-beta-dehydrogenase isozyme 1; Short Chain Dehydrogenase/Reductase Family 26C,Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8566