Learn More
Biomatik Corporation Mouse 17-Beta-Hydroxysteroid Dehydrogenase Type 10 (HSD17b10) ELISA Kit
Supplier:  Biomatik Corporation EKU02008
Encompass_Preferred
Mouse 17-Beta-Hydroxysteroid Dehydrogenase Type 10 (HSD17b10) ELISA Kit for detecting HSD17b10 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O08756. Target Synonyms: 17b-HSD10; ABAD; ERAB; HADH2; HCD2; MHBD; SCHAD; SDR5C1, MRPP2; CAMR; 3-Hydroxyacyl-CoA Dehydrogenase Type-2; AB-Binding Alcohol Dehydrogenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9159