Learn More
Instech Mouse 1ch magnetic Vascular Access Button tether kit for ABS: VABM1T/25, 375/25PS, 9in 015 sil (DV: 65ul)
SDP
Catalog No. NC1357879
Encompass
The Instech KVABM1T/25-ABS™ is a one-channel mouse VAB™ tether kit configured for ABS2™ automated blood sampling. It includes the VABM1T/25 tether, a 25ga 375/25PS swivel, 9in (23cm) of 0.015in (0.38mm) ID silicone tubing, 24in (60cm) PU tubing, and an LS25 luer stub. Designed for one-channel mouse VABs™, it provides a dead volume of 65µL, and the magnetic connection minimizes force for consistent ABS™ workflows.
Features
  • ABS™/ABS2™-configured 1ch mouse tether
  • 9in (23cm) 0.015in (0.38mm) silicone tubing
  • 24in (60cm) PU tubing
  • 375/25PS swivel, 25ga
  • Dead volume: 65µL
Catalog No. NC1357879 Supplier Instech Supplier No. KVABM1T/25ABS
The Instech KVABM1T/25-ABS™ is a one-channel mouse VAB™ tether kit configured for ABS2™ automated blood sampling. It includes the VABM1T/25 tether, a 25ga 375/25PS swivel, 9in (23cm) of 0.015in (0.38mm) ID silicone tubing, 24in (60cm) PU tubing, and an LS25 luer stub. Designed for one-channel mouse VABs™, it provides a dead volume of 65µL, and the magnetic connection minimizes force for consistent ABS™ workflows.
Features
  • ABS™/ABS2™-configured 1ch mouse tether
  • 9in (23cm) 0.015in (0.38mm) silicone tubing
  • 24in (60cm) PU tubing
  • 375/25PS swivel, 25ga
  • Dead volume: 65µL