Learn More
Alpha Diagnostic International Mouse IgG ELISA Kit, 96 tests, Quantitative
SDP
Supplier:  Alpha Diagnostic International 6320
Encompass
The antibodies used in this kit are not specific for any IgG subtype (IgG1, IgG2, etc.). This kit is also specific to IgG and does not show any significant reactivity to mouse IgA, IgM, IgE or other serum proteins.
Catalog No. NC1484459