Learn More
ACROBiosystems ACROBiosystems Mouse 4-1BB /TNFRSF9 Protein, Fc Tag (MALS verified)
Catalog No. 502106219
Encompass_Preferred
Recombinant Protein;1MG;Mouse 4-1BB, Fc Tag (41B-M5258) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Leu 211 (Accession # Q8R037-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.7 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Mouse 4-1BB, Fc Tag (Cat. No. 41B-M5258) at 2 μg/mL (100 μL/well) can bind Human 4-1BB Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. 41L-H5254) with a linear range of 20-312 ng/mL (QC tested).;Immobilized Mouse 4-1BB, Fc Tag (Cat. No. 41B-M5258) at 2 μg/mL (100 μL/well) can bind Biotinylated Human 4‑1BB Ligand, Fc Tag, Avi Tag (Cat. No. 41L-H82F9) with a linear range of 0.01-0.156 μg/mL (Routinely tested).
Catalog No. 50-210-6219 Supplier ACROBiosystems Supplier No. 41BM52581MG
Recombinant Protein;1MG;Mouse 4-1BB, Fc Tag (41B-M5258) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Leu 211 (Accession # Q8R037-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.7 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Mouse 4-1BB, Fc Tag (Cat. No. 41B-M5258) at 2 μg/mL (100 μL/well) can bind Human 4-1BB Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. 41L-H5254) with a linear range of 20-312 ng/mL (QC tested).;Immobilized Mouse 4-1BB, Fc Tag (Cat. No. 41B-M5258) at 2 μg/mL (100 μL/well) can bind Biotinylated Human 4‑1BB Ligand, Fc Tag, Avi Tag (Cat. No. 41L-H82F9) with a linear range of 0.01-0.156 μg/mL (Routinely tested).