Learn More
BIOHIPPO Mouse 4-HNE Microsample
SDP
Catalog No. 5000315154
Encompass
Mouse 4-HNE Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of 4-HNE in Serum plasma and other biological fluids It provides a detection range of 2 35-150 ng/mL with a sensitivity of 1 1 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse 4-HNE Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse 4-HNE Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse 4-HNE biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15154 Supplier BIOHIPPO Supplier No. BHE15210705
Mouse 4-HNE Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of 4-HNE in Serum plasma and other biological fluids It provides a detection range of 2 35-150 ng/mL with a sensitivity of 1 1 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse 4-HNE Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse 4-HNE Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse 4-HNE biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color