Learn More
American Research Products Inc Mouse 4 HydroXynonenal HNE E
Catalog No. 5000329427
Encompass
Mouse 4-HydroXynonenal (HNE) ELISA Kit from CUSABIO Detection Range 15 6 pmol/mL-1000 pmol/mL Sensitivity 3 9 pmol/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29427 Supplier American Research Products Inc Supplier No. CSBE13412M
Mouse 4-HydroXynonenal (HNE) ELISA Kit from CUSABIO Detection Range 15 6 pmol/mL-1000 pmol/mL Sensitivity 3 9 pmol/mL Sample serum plasma Method Sandwich