Learn More
Sino Biological Recombinant Human 4-1BB Ligand/TNFSF9 Protein (AVI & Fc Tag), Biotinylated
Supplier:  Sino Biological 15693H42HB100
Encompass_Preferred
A DNA sequence encoding the human TNFSF9 (NP_003802.1) (Arg71-Glu254) was expressed with a N-terminal AVI tag followed by a Fc region of human IgG1 tag. The purified protein was biotinylated in vitro.
Catalog No. 50-222-2115