Learn More
BIOHIPPO Mouse a1ACT Microsample
SDP
Catalog No. 5000315309
Encompass
Mouse a1ACT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of a1ACT in serum plasma and other biological fluids It provides a detection range of 15 63-1000 g/mL with a sensitivity of 5 01 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse a1ACT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse a1ACT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse a1ACT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15309 Supplier BIOHIPPO Supplier No. BHE15212692
Mouse a1ACT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of a1ACT in serum plasma and other biological fluids It provides a detection range of 15 63-1000 g/mL with a sensitivity of 5 01 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse a1ACT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse a1ACT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse a1ACT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color