Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Monoclonal Anti-Monkeypox A29L Antibody, Human IgG1 (Clone 3H9) - 1 mg

Catalog No. 502370263
Encompass_Preferred

Recombinant chimeric monoclonal antibody (clone 3H9) directed against the A29L protein of Monkeypox virus (MPXV). Supplied lyophilized for long-term storage and validated for ELISA-based detection of A29L.

  • Size: 1 mg (500 μg x 2).
  • Clone name: 3H9.
  • Host and isotype: mouse variable region with human IgG1 kappa constant.
  • Formulation: lyophilized from 0.22 μm filtered PBS, pH 7.4, with trehalose.
  • Purification: protein A/G purified.
  • Purity: approximately 90% by SDS-PAGE and SEC-MALS.
  • Immunogen: recombinant MPXV A29L produced in human 293 cells.
  • Application: ELISA, recommended 0.5-125 ng/mL.
  • Storage: lyophilized 12 months at -20°C to -70°C; reconstituted stability 3 months at -70°C; avoid repeated freeze-thaw cycles.

Catalog No. 50-237-0263 Supplier ACROBiosystems Supplier No. A2LM5771MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant chimeric monoclonal antibody (clone 3H9) directed against the A29L protein of Monkeypox virus (MPXV). Supplied lyophilized for long-term storage and validated for ELISA-based detection of A29L.

  • Size: 1 mg (500 μg x 2).
  • Clone name: 3H9.
  • Host and isotype: mouse variable region with human IgG1 kappa constant.
  • Formulation: lyophilized from 0.22 μm filtered PBS, pH 7.4, with trehalose.
  • Purification: protein A/G purified.
  • Purity: approximately 90% by SDS-PAGE and SEC-MALS.
  • Immunogen: recombinant MPXV A29L produced in human 293 cells.
  • Application: ELISA, recommended 0.5-125 ng/mL.
  • Storage: lyophilized 12 months at -20°C to -70°C; reconstituted stability 3 months at -70°C; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.