Learn More
Sino Biological Recombinant Mouse ACE2 Protein (His & hFc Tag)
Supplier:  Sino Biological 50249M03H1
Encompass_Preferred
A DNA sequence encoding the mouse ACE2 isoform 1 (Q8R0I0-1) extracellular domain (Met 1-Thr 740) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-3336