Learn More
Sino Biological Recombinant Mouse ACE2 Protein (His Tag)
Supplier:  Sino Biological 50249M08H1
Encompass_Preferred
A DNA sequence encoding the mouse ACE2 isoform 1 (Q8R0I0-1) extracellular domain (Met 1-Thr 740) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3334