Learn More
American Research Products Inc Mouse Acetylcholinesterase AC
Catalog No. 5000328526
Encompass
Mouse Acetylcholinesterase (AChE) ELISA Kit from CUSABIO Detection Range 0 312 mU/mL-20 mU/mL Sensitivity 0 078 mU/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28526 Supplier American Research Products Inc Supplier No. CSBE17521M
Mouse Acetylcholinesterase (AChE) ELISA Kit from CUSABIO Detection Range 0 312 mU/mL-20 mU/mL Sensitivity 0 078 mU/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich