Learn More
American Research Products Inc Mouse acidic fibroblast growth
Catalog No. 5000329595
Encompass
Mouse acidic fibroblast growth factor aFGF/FGF-1 ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29595 Supplier American Research Products Inc Supplier No. CSBE07327M
Mouse acidic fibroblast growth factor aFGF/FGF-1 ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich