Learn More
Vector Laboratories Mouse ACLP / AEBP1 (Sandwich ELISA) ELISA Kit, 1 plate
SDP
Supplier:  Vector Laboratories LSF107631
Encompass
96-well enzyme-linked immunosorbent assay (ELISA) for the Quantitative detection of Mouse ACLP / AEBP1 in samples of Plasma and Serum. It is based upon a Sandwich assay principle and can be used to detect levels of ACLP / AEBP1 as low as 78 picograms per milliliter.
Catalog No. NC1853734