Learn More
Sino Biological Recombinant Anti-Prostatic Acid Phosphatase/ACPP Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 51018R01920
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Prostatic Acid Phosphatase/ACPP (rM Prostatic Acid Phosphatase/ACPP; Catalog#51018-M08H; Q8CE08-1; Met1-Arg381).
Catalog No. 50-255-3029