Learn More
Sino Biological Anti-Prostatic Acid Phosphatase/ACPP Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51018T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Prostatic Acid Phosphatase / ACPP (Catalog#51018-M08H; Q8CE08-1; Met1-Arg381). Prostatic Acid Phosphatase / ACPP specific IgG was purified by Mouse Prostatic Acid Phosphatase / ACPP affinity chromatography.
Catalog No. 50-255-3026