Learn More
American Research Products Inc Mouse actiVated protein C APC
Catalog No. 5000326232
Encompass
Mouse actiVated protein C APC ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26232 Supplier American Research Products Inc Supplier No. CSBE09914M
Mouse actiVated protein C APC ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich