Learn More
Cell Signaling Technology Mouse Activated T Cell Markers Flow Cytometry Panel, Mouse, 200 Assays, 1 Kit
SDP
Catalog No. 502732495
Encompass
The Mouse Activated T Cell Markers Flow Cytometry Panel is designed to identify activated mouse T cells. This kit includes monoclonal antibodies for immunophenotyping T cell subsets and monitoring activation states. T cells are identified by CD3 expression, with helper (CD4+) and cytotoxic (CD8+) subsets defined by their respective co-receptor expression. Activation is determined by upregulated CD69 and IL-2Rα/CD25 markers, which are also constitutively expressed on regulatory T cells. Antibodies are purified via affinity chromatography and are compatible with single staining mix protocols.
  • Enables identification of CD3+CD4+ helper and CD3+CD8+ cytotoxic T cells
  • Monitors activation markers CD69 and IL-2Rα/CD25
  • Compatible with live cell flow cytometry
  • Purified via affinity chromatography
  • Supplied in 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2
Catalog No. 50-273-2495 Supplier Cell Signaling Technology Supplier No. 62447S
The Mouse Activated T Cell Markers Flow Cytometry Panel is designed to identify activated mouse T cells. This kit includes monoclonal antibodies for immunophenotyping T cell subsets and monitoring activation states. T cells are identified by CD3 expression, with helper (CD4+) and cytotoxic (CD8+) subsets defined by their respective co-receptor expression. Activation is determined by upregulated CD69 and IL-2Rα/CD25 markers, which are also constitutively expressed on regulatory T cells. Antibodies are purified via affinity chromatography and are compatible with single staining mix protocols.
  • Enables identification of CD3+CD4+ helper and CD3+CD8+ cytotoxic T cells
  • Monitors activation markers CD69 and IL-2Rα/CD25
  • Compatible with live cell flow cytometry
  • Purified via affinity chromatography
  • Supplied in 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2