Learn More
Biomatik Corporation Mouse Activating Transcription Factor 6 (ATF6) ELISA Kit
Supplier:  Biomatik Corporation EKU02108
Encompass_Preferred
Mouse Activating Transcription Factor 6 (ATF6) ELISA Kit for detecting ATF6 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.126ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F6VAN0. Gene ID: 226641. Target Synonyms: Processed cyclic AMP-dependent transcription factor ATF-6 alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9154