Learn More
Crystal Chem Inc Mouse Active GIP ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 81511
Encompass
The Mouse Active GIP ELISA is a highly sensitive assay that is used to quantify levels of mouse GIP active in plasma and cell culture. Highlights: Kits use a sample volume of only 15 µL. High Sensitivity of 1.56 pM. Dynamic range of 1.56 - 100 pM. Highly precise with low CV's. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7897