Learn More
Raybiotech Inc Mouse Activin A ELISA Kit
Supplier:  Raybiotech Inc ELMACTIVINA1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.5 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse Activin A.
Catalog No. 50-194-9919