Learn More
Sino Biological Recombinant Anti-ACVR2B Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50173R105100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ACVR2B (rM ACVR2B ; Catalog#50173-M08H; NP_031423.1; Met1-Thr134).
Catalog No. 50-240-7922