Learn More
Sino Biological Anti-ACVR2B Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50173RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse ACVR2B extracellular domain (rM ACVR2B; Catalog#50173-M08H; NP_031423.1; Met 1-Thr 134). ACVR2B specific IgG was purified by mouse ACVR2B affinity chromatography.
Catalog No. 50-240-8403