Learn More
Sino Biological Anti-ADAM15 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51001RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse ADAM15 (Catalog#51001-M08H; O88839-1; Met1-Thr697). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3038