Learn More
Sino Biological Anti-ADAM15 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51001RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse ADAM15 (rM ADAM15; Catalog#51001-M08H; O88839-1; Met 1-Thr 697). ADAM15 specific IgG was purified by Mouse ADAM15 affinity chromatography.
Catalog No. 50-255-3037