Learn More
Sino Biological Mouse ADAM9 Protein (His Tag) 500ug
Supplier:  Sino Biological 50044M08H500
Encompass_Preferred
A DNA sequence encoding the mouse ADAM9 (Q61072) extracellular domain (Met 1-Asp 697) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-6794