Learn More
American Research Products Inc Mouse ADAMTS like protein 4 A
Catalog No. 5000327548
Encompass
Mouse ADAMTS-like protein 4 (ADAMTSL4) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27548 Supplier American Research Products Inc Supplier No. CSBEL001320MO
Mouse ADAMTS-like protein 4 (ADAMTSL4) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich