Learn More
Raybiotech Inc Mouse ADAMTS-1 (METH1) ELISA Kit
Supplier:  Raybiotech Inc ELMADAMTS11
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.4 ng/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects mouse and human ADAMTS1. Other species not determined.
Catalog No. 50-194-9920