Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse ADAMTS1 ELISA Kit

Catalog No. p-7219806
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EM4RBX5 5 x 96 Tests
EM4RB 96 Tests
EM4RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EM4RBX5 Supplier Invitrogen™ Supplier No. EM4RBX5
Only null left
Add to Cart
Add to Cart

ELISA

Mouse ADAMTS1 quantitates mouse ADAMTS1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse ADAMTS1.

ADAMTS-1 is a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motif) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene contains two disintegrin loops and three C-terminal TS motifs and has anti-angiogenic activity. The expression of this gene may be associated with various inflammatory processes as well as development of cancer cachexia. This gene is likely to be necessary for normal growth, fertility, and organ morphology and function.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 1; a disintegrin and metalloproteinase with thrombospondin motifs 1 (ADAMTS-1); a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 1; a disintegrin-like and metallopeptidse (reprolysin type) with thrombospondin type 1 motif, 1; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 1; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 1; ADAM metallopeptidase with thrombospondin type 1 motif, 1; ADAMs; ADAMTS; ADAM-TS 1; Adamts1; ADAM-TS1; ADAMTS-1; C3-C5; human metalloproteinase with thrombospondin type 1 motifs; KIAA1346; metalloendopeptidases; METH1; METH-1
Assay Range 0.4 to 100 ng/mL
Assay Sensitivity 0.4 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 11504
Gene Symbol ADAMTS1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target ADAMTS1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.