Learn More
World Precision Instrument Mouse Adapter with 45 Deg ear bars
SDP
Catalog No. 502539069
Encompass
The Mouse Adaptor holds the mouse's head by means of a palate bar and nose clamp, without ear bars. Head angle may be adjusted to achieve a flat skull or other approach angles.
Catalog No. 50-253-9069 Supplier World Precision Instrument Supplier No. 505391
The Mouse Adaptor holds the mouse's head by means of a palate bar and nose clamp, without ear bars. Head angle may be adjusted to achieve a flat skull or other approach angles.