Learn More
World Precision Instrument Mouse Adaptor
SDP
Catalog No. 502539173
Encompass
  • The mouse's head by means of a palate bar and nose clamp, without ear bars. Head angle may be adjusted to achieve a flat skull or other approach angles.
  • The mouse head by means of a palate bar and nose clamp, without ear bars. (Ear bars can cause breathing difficulties and bleeding in mice.) Head angle may be adjusted to achieve a flat skull or other approach angles. Fits 505211, 505293,505326 and 505351 series stereotaxic instruments.
Catalog No. 50-253-9173 Supplier World Precision Instrument Supplier No. 505242
  • The mouse's head by means of a palate bar and nose clamp, without ear bars. Head angle may be adjusted to achieve a flat skull or other approach angles.
  • The mouse head by means of a palate bar and nose clamp, without ear bars. (Ear bars can cause breathing difficulties and bleeding in mice.) Head angle may be adjusted to achieve a flat skull or other approach angles. Fits 505211, 505293,505326 and 505351 series stereotaxic instruments.