Learn More
RWD LIFE SCIENCE INC MOUSE ADAPTOR
SDP
Catalog No. NC3388877
Encompass
It holds the mouse head by a palate and incisor, and offers vertical regulation to make the head level or at certain angle. The Vertical adjustable range from +10mm to -20mm, with 100μm resolution; Horizontal adjustable range of 43.5mm; Horizontal rotatable range of 35°.
Catalog No. NC3388877 Supplier RWD LIFE SCIENCE INC Supplier No. 68055EA
It holds the mouse head by a palate and incisor, and offers vertical regulation to make the head level or at certain angle. The Vertical adjustable range from +10mm to -20mm, with 100μm resolution; Horizontal adjustable range of 43.5mm; Horizontal rotatable range of 35°.